missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCMA/TNFRSF17 Antibody (CD269/8508R), Novus Biologicals™
Rabbit Monoclonal Antibody
£246.00 - £446.00
Specifications
| Antigen | BCMA/TNFRSF17 |
|---|---|
| Clone | CD269/8508R |
| Concentration | 0.2 mg/ml |
| Dilution | ELISA, Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | ELISA, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18608124
|
Novus Biologicals
NBP3-20435-20ug |
20 μg |
£246.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18655534
|
Novus Biologicals
NBP3-20435-100ug |
100 μg |
£446.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BCMA/TNFRSF17 Monoclonal antibody specifically detects BCMA/TNFRSF17 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
| BCMA/TNFRSF17 | |
| 0.2 mg/ml | |
| ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Signal Transduction | |
| 10mM PBS with 0.05% BSA | |
| 608 | |
| IgG κ | |
| Protein A or G purified |
| CD269/8508R | |
| ELISA, Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| B cell maturation antigen, B-cell maturation protein, BCMAtumor necrosis factor receptor superfamily member 17, BCMB-cell maturation factor, CD269, CD269 antigen, tumor necrosis factor receptor superfamily, member 17 | |
| Recombinant human BCMA/TNFRSF17 protein fragment (around aa 1-184) (exact sequence is proprietary) | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title