missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems BCMA protein-coupled magnetic beads

Product Code. 17196481
Click to view available options
Quantity:
2 mg
10 mg
Unit Size:
10mg
2mg

Product Code. 17196481

Brand: ACROBiosystems MBSK00410mg Beads

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

For capturing the anti-BCMA antibody from various assay systems

The biotinylated BCMA protein was conjugated to streptavidin magnetic beads. This pre-coupled magnetic bead product can capture the anti-BCMA antibody from various assay systems. The beads are in uniform size, narrow size distribution with large surface area and unique surface coating, which can help you get the best performance and highly reproducible results. This Human BCMA-coupled magnetic beads will bring great convenience with minimum non-specific binding and developed protocols. This ready-to-use product could greatly save your time and hassle.

Specifications

Kit Contents The conjugation was achieved by means of the binding between streptavidin and biotin. Streptavidin (SA) has an extraordinarily high affinity for biotin with a dissociation constant (Kd) on the order of 10-14 mol/L Thus, the binding of streptavidin and bi
Content And Storage -20°C, Avoid light
Shipping Condition RT
Quantity 10 mg
Type Magnetic Affinity Bead
Species Human
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.