missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems BCMA protein-coupled magnetic beads
Click to view available options
Quantity:
2 mg
10 mg
Unit Size:
10mg
2mg
Description
The biotinylated BCMA protein was conjugated to streptavidin magnetic beads. This pre-coupled magnetic bead product can capture the anti-BCMA antibody from various assay systems. The beads are in uniform size, narrow size distribution with large surface area and unique surface coating, which can help you get the best performance and highly reproducible results. This Human BCMA-coupled magnetic beads will bring great convenience with minimum non-specific binding and developed protocols. This ready-to-use product could greatly save your time and hassle.
Specifications
Specifications
| Kit Contents | The conjugation was achieved by means of the binding between streptavidin and biotin. Streptavidin (SA) has an extraordinarily high affinity for biotin with a dissociation constant (Kd) on the order of 10-14 mol/L Thus, the binding of streptavidin and bi |
| Content And Storage | -20°C, Avoid light |
| Shipping Condition | RT |
| Quantity | 2 mg |
| Type | Magnetic Affinity Bead |
| Species | Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction