missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCL9L Polyclonal Antibody

BCL9L Polyclonal Antibody
Brand: Thermo Scientific PA5105738
This item is not returnable.
View return policy
Description
BCL9L Polyclonal Antibody for Western Blot
Specifications
| BCL9L | |
| Polyclonal | |
| Unconjugated | |
| BCL9L | |
| B cell CLL/lymphoma 9-like; B9L; BC003321; B-cell CLL/lymphoma 9-like; B-cell CLL/lymphoma 9-like protein; B-cell lymphoma 9-like protein; BCL9-2; BCL9L; BCL9-like protein; BCL9-related beta-catenin-binding protein; DLNB11; nuclear co-factor of beta-catenin signalling; protein BCL9-2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 283149 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q86UU0 | |
| BCL9L | |
| A synthesized peptide derived from human BCL9L(Accession Q86UU0), corresponding to amino acid residues M1-T45. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction