missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCL9 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22947-1-AP-150UL
This item is not returnable.
View return policy
Description
BCL9 is involved in signal transduction through the Wnt pathway. It promotes beta-catenin′s transcriptional activity. BCL9 is associated with B-cell acute lymphoblastic leukemia. The overexpression of BCL9 may be of pathogenic significance in B-cell maligciesSpecifications
| BCL9 | |
| Polyclonal | |
| Unconjugated | |
| BCL9 | |
| B cell CLL/lymphoma 9, B cell CLL/lymphoma 9 protein, B cell lymphoma 9 protein, BCL9, LGS, Protein legless homolog | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 607 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.44 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O00512 | |
| BCL9 | |
| BCL9 Fusion Protein Ag19158 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction