missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCL9, Mouse anti-Human, Clone: 1A2, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00000607-M02.100ug
This item is not returnable.
View return policy
Description
Sequence: DDDSPPARSPNLPSMNNMPGMGINTQNPRISGPNPVVPMPTLSPMGMTQPLSHSNQMPSPNAVGPNIPPHGVPMGPGLMSHNPIMGHGSQEPPMVPQGRMSpecifications
| BCL9 | |
| Monoclonal | |
| Unconjugated | |
| B-cell CLL/lymphoma 9 | |
| LGS/MGC131591 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Western Blot | |
| 1A2 | |
| In 1x PBS, pH 7.4 | |
| NM_004326 | |
| BCL9 | |
| BCL9 (NP_004317, 1036 a.a. ∼ 1135 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 607 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction