missing translation for 'onlineSavingsMsg'
Learn More

BCL9-2 Antibody, DyLight 594, Novus Biologicals™

Product Code. 18430287 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18430287 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18430287 Supplier Novus Biologicals Supplier No. NBP176542DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

BCL9-2 Polyclonal antibody specifically detects BCL9-2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen BCL9-2
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias B-cell CLL/lymphoma 9-like, B-cell CLL/lymphoma 9-like protein, B-cell lymphoma 9-like protein, BCL9-like protein, DLNB11BCL9-2, nuclear co-factor of beta-catenin signalling, Protein BCL9-2
Host Species Rabbit
Immunogen Antibody was raised against a 20 amino acid synthetic peptide near the amino terminus of human Bcl9L. The immunogen is located within amino acids 20 - 70 of Bcl9L. Amino Acid Squence: PAKPMHPENKLTNHGKTGNG
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 283149
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.