missing translation for 'onlineSavingsMsg'
Learn More

BCL7B Antibody (OTI4B10) [Alexa Fluor™ 594], Novus Biologicals™

Product Code. 30687939 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30687939 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30687939 Supplier Novus Biologicals Supplier No. NBP344964AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

BCL7B Monoclonal antibody specifically detects BCL7B in Human,Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen BCL7B
Applications Western Blot
Classification Monoclonal
Clone OTI4B10
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias Allergen Hom s 3, B-cell CLL/lymphoma 7 protein family member B, B-cell CLL/lymphoma 7B
Host Species Mouse
Immunogen Full length human recombinant protein of human BCL7B (NP_001698) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9275
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.