missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BCL7A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582696
This item is not returnable.
View return policy
Description
Immunogen sequence: QADGKEHPGA EDASDEQNSQ SSMEHSMNSS EKVDRQPSGD SGLAAETSAI SQDLEGVPPS KKMKLEASQQ NSE Highest antigen sequence indentity to the following orthologs: Rat - 90%, Mouse - 89%.
This gene is directly involved, with Myc and IgH, in a three-way gene translocation in a Burkitt lymphoma cell line. As a result of the gene translocation, the N-terminal region of the gene product is disrupted, which is thought to be related to the pathogenesis of a subset of high-grade B cell non-Hodgkin lymphoma. The N-terminal segment involved in the translocation includes the region that shares a strong sequence similarity with those of BCL7B and BCL7C. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| BCL7A | |
| Polyclonal | |
| Unconjugated | |
| BCL7A | |
| 4432415N06Rik; AI448316; B cell CLL/lymphoma 7A; B-cell CLL/lymphoma 7 protein family member A; B-cell CLL/lymphoma 7A; B-cell CLL/lymphoma-7; BCL 7; BCL tumor suppressor 7A; BCL7; Bcl7a; LOC690085; similar to B-cell CLL/lymphoma 7A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 605 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry, Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q4VC05 | |
| BCL7A | |
| Recombinant protein corresponding to Human BCL7A. Recombinant protein control fragment (Product #RP-92559). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction