Learn More
BCL6 Monoclonal Antibody (GI191E), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen 14-9887-80
Description
Description: This GI191E monoclonal antibody reacts with mouse and human Bcl-6, which is also known as B-cell lymphoma 6. A member of the POZ family of transcriptional repressors, Bcl-6 is expressed most abundantly in germinal center B cells, as well as in differentiating and activated B cells. This 79-kDa protein is essential for germinal center B cell proliferation and survival. Moreover, during B cell development, Bcl-6 plays an important role in promoting pre-B cell survival to enable the generation of a diverse B cell repertoire. Moreover, Bcl-6 is expressed constitutively in diffuse large B cell lymphomas (DLBCLs), Burkitt's lymphoma, and non-Hodgkin's lymphoma. Reports also suggest expression of Bcl-6 in CD4+ T follicular helper cells and memory T cells. Bcl-6 has been reported to be antagonized by Blimp-1. Applications Reported: This GI191E antibody has been reported for use in immunoblotting (WB) and immunohistochemistry (FFPE). Applications Tested: This GI191E antibody has been tested by immunoblotting of reduced cell lysate prepared from A20 cells and by immunohistochemistry on human tissue using high pH antigen retrieval. This antibody can be used at less than or equal to 5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
2 μm post-manufacturing filtered. Bcl-6 proto-oncogene product (a Kruppel-type zinc-finger protein) is mainly expressed in normal germinal center B cells and related lymphomas. Bcl-6 is involved in chromosome rearrangements at 3q27 in non-Hodgkin′s lymphomas and Bcl-6 rearrangements have been detected in 33%-45% of diffuse large B cell lymphomas. Bcl-6 has been detected immunohistochemically in follicular lymphomas, diffuse large B cell lymphomas, Burkitt′s lymphomas and in nodular, lymphocyte predomit Hodgkin′s disease.
Specifications
| BCL6 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| P41182, P41183 | |
| Bcl6 | |
| Affinity chromatography | |
| RUO | |
| 12053, 604 | |
| 4° C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| GI191E | |
| Unconjugated | |
| BCL6 | |
| B-cell lymphoma 6 protein; BCL-6; Zinc finger protein 51; LAZ-3 protein; BCL-5; Zinc finger and BTB domain-containing protein 27; ZNF51; ZBTB27; LAZ3; BCL5; BCL6 | |
| Mouse | |
| 25 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.