Learn More
BCL3 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-23959-100UL
Description
The BCL3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human BCL3. This antibody recognizes human, mouse antigen. The BCL3 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| BCL3 | |
| 1000 μg/mL | |
| B cell CLL/lymphoma 3, B cell lymphoma 3 protein, BCL3, D19S37, Proto oncogene BCL3 | |
| 100 μL | |
| 602 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |