missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BCL2L15 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114306
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
BCL2L15 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Proteins of the Bcl-2 family are critical regulators of apoptosis. Proapoptotic members, like Bax, contain three of the four Bcl-2 homology regions (BH1-3), while BH3-only proteins, like Bim, possess only the short BH3 motif. Database searches revealed Bfk, an unusual novel member of the Bcl-2 family that contains a BH2 and BH3 region but not BH1 or BH4. Bfk is thus most closely related to Bcl-G(L). It lacks a C-terminal membrane anchor and is cytosolic. Enforced expression of Bfk weakly promoted apoptosis and antagonized Bcl-2's prosurvival function. Like Bcl-G(L), Bfk did not bind to any Bcl-2 family members, even though its BH3 motif can mediate association with prosurvival proteins. Low amounts of Bfk were found in stomach, ovary, bone marrow and spleen, but its level in the mammary gland rose markedly during pregnancy, suggesting that Bfk may play a role in mammary development.
Specifikationer
| BCL2L15 | |
| Polyclonal | |
| Unconjugated | |
| BCL2L15 | |
| BCL2 like 15; BCL2L15; bcl2-L-15; BCL2-like 15; bcl-2-like protein 15; BCLl2-like 15; Bfk; C1orf178; Gm566; pro-apoptotic Bcl-2 protein; RGD1562344 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 229672, 295337, 440603 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q08ED0, Q5TBC7 | |
| BCL2L15 | |
| Recombinant protein within Human BCL2L15 aa 1-163. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering