missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BCL2L15 Monoclonal Antibody (K06343_5E8), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA555946
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: TECIVNTLLM DFLSPTLQVA SRNLCCVDEV DSGEPCSFDV AIIAGRLRML GDQFNGELEA SAKNVIAETI KGQTGAILQD TVESLSKTWC AQDSSLAYER AFLAVSVKLL EYMAHIAPEV VGQVAIPMTG MINGNQAIRE FIQGQGGWEN
Proteins of the Bcl-2 family are critical regulators of apoptosis. Proapoptotic members, like Bax, contain three of the four Bcl-2 homology regions (BH1-3), while BH3-only proteins, like Bim, possess only the short BH3 motif. Database searches revealed Bfk, an unusual novel member of the Bcl-2 family that contains a BH2 and BH3 region but not BH1 or BH4. Bfk is thus most closely related to Bcl-G(L). It lacks a C-terminal membrane anchor and is cytosolic. Enforced expression of Bfk weakly promoted apoptosis and antagonized Bcl-2's prosurvival function. Like Bcl-G(L), Bfk did not bind to any Bcl-2 family members, even though its BH3 motif can mediate association with prosurvival proteins. Low amounts of Bfk were found in stomach, ovary, bone marrow and spleen, but its level in the mammary gland rose markedly during pregnancy, suggesting that Bfk may play a role in mammary development.
Specifications
| BCL2L15 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q5TBC7 | |
| BCL2L15 | |
| Recombinant human BCL2L15 (immunogen range is 11-160). | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| K06343_5E8 | |
| Unconjugated | |
| BCL2L15 | |
| BCL2 like 15; BCL2L15; bcl2-L-15; BCL2-like 15; bcl-2-like protein 15; BCLl2-like 15; Bfk; C1orf178; Gm566; pro-apoptotic Bcl-2 protein; RGD1562344 | |
| Mouse | |
| Protein A | |
| RUO | |
| 440603 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction