missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCL2L15 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33444-20ul
This item is not returnable.
View return policy
Description
BCL2L15 Monoclonal antibody specifically detects BCL2L15 in Human samples. It is validated for ELISA,Western Blot
Specifications
| BCL2L15 | |
| Monoclonal | |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| BCL2-like 15, FLJ22588, pro-apoptotic Bcl-2 protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-163 of human BCL2L15 (NP_001010922.1).,, Sequence:, MKSSQTFEEQTECIVNTLLMDFLSPTLQVASRNLCCVDEVDSGEPCSFDVAIIAGRLRMLGDQFNGELEASAKNVIAETIKGQTGAILQDTVESLSKTWCAQDSSLAYERAFLAVSVKLLEYMAHIAPEVVGQVAIPMTGMINGNQAIREFIQGQGGWENLES | |
| 20 μL | |
| Apoptosis, Cell Biology | |
| 440603 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction