missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BCL2L12 Recombinant Rabbit Monoclonal Antibody (23GB3695), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | BCL2L12 |
|---|---|
| Clone | 23GB3695 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088627
![]() |
Invitrogen™
MA553636 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088920
|
Invitrogen™
MA553637 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BCL2L12 Recombinant Monoclonal antibody specifically detects BCL2L12 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| BCL2L12 | |
| Flow Cytometry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9HB09 | |
| 83596 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB3695 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 2810475P17Rik; 5430429M05Rik; BCL2 like 12; Bcl-2 related proline-rich protein; Bcl2l12; Bcl2-L12; Bcl2-L-12; BCL2-like 12 (proline rich); bcl-2-like protein 12; Bcl-2-related proline-rich protein; Bcl-L12; BPR | |
| A synthetic peptide of human BCL2L12 (250-300AA). | |
| Primary | |
| -20°C | |
| BCL2L12 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title