missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ BCL2L10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100704
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
Antibody detects endogenous levels of total BCL2L10.
The protein encoded by this gene belongs to the BCL-2 protein family. BCL-2 family members form hetero- or homodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. The protein encoded by this gene contains conserved BH4, BH1 and BH2 domains. This protein can interact with other members of BCL-2 protein family including BCL2, BCL2L1/BCL-X(L), and BAX. Overexpression of this gene has been shown to suppress cell apoptosis possibly through the prevention of cytochrome C release from the mitochondria, and thus activating caspase-3 activation. The mouse counterpart of this protein is found to interact with Apaf1 and forms a protein complex with Caspase 9, which suggests the involvement of this protein in APAF1 and CASPASE 9 related apoptotic pathway.
Tekniske data
| BCL2L10 | |
| Polyclonal | |
| Unconjugated | |
| BCL2L10 | |
| AA420380; anti-apoptotic protein Boo; Anti-apoptotic protein NrH; apoptosis regulator Bcl-B; AU023065; Bcl-2 homolog Diva; BCL2 like 10; BCL2L10; bcl2-L-10; Bcl2-like 10; BCL2-like 10 (apoptosis facilitator); bcl-2-like protein 10; BCLB; BCL-B; Boo; C85687; death inducer binding to vBcl-2 and Apaf-1; Diva | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10017, 12049 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9HD36, Q9Z0F3 | |
| BCL2L10 | |
| A synthesized peptide derived from human BCL2L10(Accession Q9HD36), corresponding to amino acid residues G122-Q172. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion