missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCL2L10 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
£429.00
Specifications
| Antigen | BCL2L10 |
|---|---|
| Applications | Western Blot, ELISA, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | PerCP |
| Form | Purified |
Description
BCL2L10 Polyclonal antibody specifically detects BCL2L10 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
| BCL2L10 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Apoptosis regulator Bcl-B, apoptotic protein NrH, bcl2-L-10, BCL2-like 10 (apoptosis facilitator), bcl-2-like protein 10, BCLB, BCL-B, Boo, Diva, MGC129810, MGC129811 | |
| Antibody was raised against a 16 amino acid synthetic peptide from near the amino terminus of human Bcl-B. The immunogen is located within amino acids 90 - 140 of Bcl-B. Amino Acid Squence: RWKKWGFQPRLKEQEGD | |
| Primary | |
| Store at 4°C in the dark. |
| Western Blot, ELISA, Immunocytochemistry | |
| PerCP | |
| Rabbit | |
| Apoptosis | |
| PBS | |
| 10017 | |
| IgG | |
| Peptide affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title