missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BCL2A1 Recombinant Rabbit Monoclonal Antibody (23GB4820), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552757
This item is not returnable.
View return policy
Description
Bcl-2 related Protein A1 is a member of the BCL-2 protein family. The proteins of this family form hetero- or homodimers and act as anti- and pro-apoptotic regulators that are involved in a wide variety of cellular activities such as embryonic development, homeostasis and tumorigenesis. The protein encoded by this gene is able to reduce the release of pro-apoptotic cytochrome c from mitochondria and block caspase activation. This gene is a direct transcription target of NF-kappa B in response to inflammatory mediators, and has been shown to be up-regulated by different extracellular signals, such as granulocyte-macrophage colony-stimulating factor (GM-CSF), CD40, phorbol ester and inflammatory cytokine TNF and IL-1, which suggests a cytoprotective function that is essential for lymphocyte activation as well as cell survival.
Specifications
| BCL2A1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q16548 | |
| BCL2A1 | |
| A synthesized peptide derived from human BCL2A1 (1-60AA). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB4820 | |
| Unconjugated | |
| BCL2A1 | |
| A1; A1-A; ACC1; ACC-1; ACC2; ACC-2; B cell leukemia/lymphoma 2 related protein A1a; BB218357; B-cell leukemia/lymphoma 2 related protein A1; B-cell leukemia/lymphoma 2 related protein A1a; B-cell leukemia/lymphoma 2 related protein A1d; BCL2 related protein A1; Bcl2a1; Bcl2a1a; Bcl2a1d; BCL2L5; bcl2-L-5; Bcl-2-like protein 5; bcl-2-related protein A1; BCL2-related protein A1; BCL2-related protein A1d; Bfl1; Bfl-1; GRS; GRS protein; HBPA1; hematopoietic BCL2-related protein A1; hemopoietic-specific early response protein; Protein BFL-1; Protein GRS | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 597 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction