missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BCL11B Monoclonal Antibody (1F8G8)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA531646
This item is not returnable.
View return policy
Description
MA5-31646 has been tested in indirect ELISA.
This gene encodes a C2H2-type zinc finger protein and is closely related to BCL11A, a gene whose translocation may be associated with B-cell malignancies. The specific function of this gene has not yet been determined. Two alternatively spliced transcript variants, which encode distinct isoforms, have been reported.
Specifications
| BCL11B | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| Q9C0K0 | |
| BCL11B | |
| Purified recombinant fragment of human BCL11B (AA: 1-150) expressed in E. coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1F8G8 | |
| Unconjugated | |
| BCL11B | |
| 9130430L19Rik; AI604821; ATL1; ATL1-alpha; ATL1-beta; ATL1-delta; ATL1-gamma; B cell CLL/lymphoma 11B; B cell leukemia/lymphoma 11B; B630002E05Rik; BAF chromatin remodeling complex subunit BCL11B; B-cell CLL/lymphoma 11B; B-cell CLL/lymphoma 11B (zinc finger protein); B-cell CLL/lymphoma 11B/T-cell receptor delta constant region fusion protein; B-cell leukemia/lymphoma 11B; B-cell lymphoma/leukaemia 11B; B-cell lymphoma/leukemia 11B; B-cell lymphoma/leukemia 11B (predicted), 3 prime; B-cell lymphoma/leukemia 11B (predicted), 5 prime; BCL11B; BCL-11B; BCL11B, BAF complex component; BCL11B/TRDC fusion; COUP-TF interacting protein 2; COUP-TF-interacting protein 2; COUP-TF-interacting protein 2 long form; CTIP2; CTIP-2; hRit1; hRIT1-alpha; LOW QUALITY PROTEIN: B-cell lymphoma/leukemia 11B; mRit1; Radiation-induced tumor suppressor gene 1 protein; Rit1; zinc f; zinc finger protein hRit1 alpha; ZNF856B | |
| Mouse | |
| Protein G | |
| RUO | |
| 64919 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction