missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCL11B Antibody (CL6426), Novus Biologicals™
Mouse Monoclonal Antibody
£328.00 - £469.00
Specifications
| Antigen | BCL11B |
|---|---|
| Clone | CL6426 |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18669440
|
Novus Biologicals
NBP2-76493-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18307050
|
Novus Biologicals
NBP2-76493 |
100 μL |
£469.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BCL11B Monoclonal specifically detects BCL11B in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| BCL11B | |
| Monoclonal | |
| Mouse | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide | |
| 64919 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: EIGIQVTPDEDDHLLSPTK | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| CL6426 | |
| Unconjugated | |
| Human, Mouse | |
| B-cell CLL/lymphoma 11B, B-cell CLL/lymphoma 11B (zinc finger protein), B-cell CLL/lymphoma 11B/T-cell receptor delta constant region fusion protein, B-cell lymphoma/leukaemia 11B, B-cell lymphoma/leukemia 11B, BCL-11B, BCL11B/TRDC fusion, COUP-TF-interacting protein 2, CTIP-2, CTIP2ZNF856B, hRit1, hRIT1-alpha, Radiation-induced tumor suppressor gene 1 protein, RIT1, zinc finger protein hRit1 alpha | |
| BCL11B | |
| IgG1 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title