missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCL11B Antibody (1F8H9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£161.00 - £435.00
Specifications
| Antigen | BCL11B |
|---|---|
| Clone | 1F8H9 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 |
| Applications | Western Blot, Flow Cytometry, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18251901
|
Novus Biologicals
NBP2-61700 |
0.1 mL |
£435.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18684337
|
Novus Biologicals
NBP2-61700-0.025ml |
0.025 mL |
£161.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BCL11B Monoclonal specifically detects BCL11B in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
| BCL11B | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA | |
| Unconjugated | |
| Mouse | |
| B-cell CLL/lymphoma 11B, B-cell CLL/lymphoma 11B (zinc finger protein), B-cell CLL/lymphoma 11B/T-cell receptor delta constant region fusion protein, B-cell lymphoma/leukaemia 11B, B-cell lymphoma/leukemia 11B, BCL-11B, BCL11B/TRDC fusion, COUP-TF-interacting protein 2, CTIP-2, CTIP2ZNF856B, hRit1, hRIT1-alpha, Radiation-induced tumor suppressor gene 1 protein, RIT1, zinc finger protein hRit1 alpha | |
| BCL11B | |
| IgG1 | |
| Protein G purified |
| 1F8H9 | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
| Monoclonal | |
| Purified | |
| Apoptosis, Cancer, Transcription Factors and Regulators, Tumor Suppressors | |
| 64919 | |
| Purified recombinant fragment of human BCL11B (AA: 1-150) expressed in E. Coli. | |
| Primary | |
| 95.5 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title