missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bcl-xL Antibody (OTI4A9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-47665
This item is not returnable.
View return policy
Description
Bcl-xL Monoclonal specifically detects Bcl-xL in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Bcl-xL | |
| Monoclonal | |
| 0.25 mg/mL | |
| Western Blot 1:200 - 1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin | |
| BCL2L1 | |
| Recombinant fragment expressed in E.coli corresponding to amino acids 1-212 of human Bcl-XL | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody is specific for Homo sapiens BCL2-like 1 (BCL2L1), nuclear gene encoding mitochondrial protein, transcript variant 1. | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OTI4A9 | |
| Unconjugated | |
| Apoptosis regulator Bcl-X, bcl2-L-1, BCL2-like 1, bcl-2-like protein 1, BCLXBCL2LBcl-X, bcl-xL, BCLXL, BCL-XL/S, bcl-xS, BCLXS, DKFZp781P2092 | |
| Mouse | |
| 25.9 kDa | |
| 0.1 mL | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 598 | |
| Human, Mouse, Rat | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction