missing translation for 'onlineSavingsMsg'
Learn More

bcl-x Mouse anti-Mouse, Rat, Clone: 2H12, Novus Biologicals™

Product Code. 18388361 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18388361 25 μg 25µg
18014764 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18388361 Supplier Bio-Techne Supplier No. NBP31188125UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

bcl-x Monoclonal antibody specifically detects bcl-x in Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen bcl-x
Applications Western Blot
Classification Monoclonal
Clone 2H12
Conjugate Unconjugated
Dilution Western Blot 1 - 5 ug/ml
Formulation PBS
Host Species Mouse
Immunogen A synthetic peptide corresponding to amino acids 3 to 14 of the human bcl-xl sequence (Uniprot: Q07817)
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 598
Target Species Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.