missing translation for 'onlineSavingsMsg'
Learn More
Learn More
bcl-x Mouse anti-Human, Clone: SPM519, Novus Biologicals™
Mouse Monoclonal Antibody
£377.00
Specifications
| Antigen | bcl-x |
|---|---|
| Clone | SPM519 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1 to 2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
Description
bcl-x Monoclonal specifically detects bcl-x in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
| bcl-x | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| 598 | |
| IgG | |
| Protein A or G purified |
| SPM519 | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| Recombinant human bcl-x protein fragment (around aa 1-135) (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title