missing translation for 'onlineSavingsMsg'
Learn More
Learn More
bcl-x Antibody (SPM337), Novus Biologicals™
Mouse Monoclonal Antibody
£211.00 - £373.00
Specifications
| Antigen | Bcl-xL |
|---|---|
| Clone | SPM337 |
| Concentration | 0.2mg/mL |
| Dilution | Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18181529
|
Novus Biologicals
NBP2-44416-0.02MG |
0.02 mg |
£211.00
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18108318
|
Novus Biologicals
NBP2-44416-0.1MG |
0.1 mg |
£373.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ensure accurate, reproducible results in Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
Bcl-xL Monoclonal specifically detects Bcl-xL in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
| Bcl-xL | |
| 0.2mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
| Apoptosis regulator Bcl-X, bcl2-L-1, BCL2-like 1, bcl-2-like protein 1, BCLXBCL2LBcl-X, bcl-xL, BCLXL, BCL-XL/S, bcl-xS, BCLXS, DKFZp781P2092 | |
| BCL2L1 | |
| IgG2a | |
| Protein A or G purified | |
| Recognizes a protein of 27kDa, identified as the Bcl-X protein. This MAb shows no cross-reaction with Bcl-2 or Bax protein. Bcl-X has two isoforms, Bcl-XL (long), a 241 amino acid protein which suppresses cell death. And Bcl-XS (short), a 178 amino acid protein lacking a 63 amino acid domain which functions as a dominant inhibitor of Bcl-2. This MAb reacts with both Bcl-XS and Bcl-XL proteins. |
| SPM337 | |
| Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Pig | |
| Q07817 | |
| 598 | |
| A synthetic peptide, aa 3-14 (Cys-QSNRELVVDFLS) of human Bcl-X protein. | |
| Primary | |
| Store at 4C. | |
| 27 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title