missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Bcl-W Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101328
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total BCL2L2.
Using a PCR-based strategy, an additional protein with life-promoting activity, designated Bcl-w, has been identified. The protein is highly conserved between mouse and human and is encoded by a gene located near the TCRalpha gene on chromosome 14. Bcl-w is expressed in myeloid cell lines but not in T and B lymphocytes, and can be found in a wide range of tissues. An alternative splicing event in exon 4 results in two transcripts. The first, Bcl-w, encodes a protein of 193 amino acids, and the second, Bcl-w/rox, encodes a protein 333 amino acids in length. The rox portion of Bcl-w/rox shows a striking 66% amino acid sequence identity with the Drosophila rox2 protein; however, the Bcl-w/rox transcript may be expressed at very low levels.
Specifications
| Bcl-W | |
| Polyclonal | |
| Unconjugated | |
| BCL2L2 | |
| Apoptosis regulator Bcl-W; AW048834; BCL2 like 2; BCL2L2; BCL2-L-2; BCL2-like 2; BCL2-like 2 protein; Bcl-2-like protein 2; Bcl2-like protein 2; BCLW; BCL-W; BCL-WEL; BCL-WS; c98; Gtrgal2; Gtrosa41; Kiaa0271; PPP1R51; protein phosphatase 1, regulatory subunit 51 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 12050, 599, 60434 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P70345, Q92843 | |
| BCL2L2 | |
| A synthesized peptide derived from human BCL2L2(Accession Q92843), corresponding to amino acid residues V18-L68. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction