missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Bcl-W Monoclonal Antibody (90316)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561930
This item is not returnable.
View return policy
Description
Bcl-W Monoclonal Antibody for Western Blot
Using a PCR-based strategy, an additional protein with life-promoting activity, designated Bcl-w, has been identified. The protein is highly conserved between mouse and human and is encoded by a gene located near the TCRalpha gene on chromosome 14. Bcl-w is expressed in myeloid cell lines but not in T and B lymphocytes, and can be found in a wide range of tissues. An alternative splicing event in exon 4 results in two transcripts. The first, Bcl-w, encodes a protein of 193 amino acids, and the second, Bcl-w/rox, encodes a protein 333 amino acids in length. The 'rox' portion of Bcl-w/rox shows a striking 66% amino acid sequence identity with the Drosophila rox2 protein; however, the Bcl-w/rox transcript may be expressed at very low levels.
Specifications
| Bcl-W | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q92843 | |
| BCL2L2 | |
| E. coli-derived recombinant human Bcl-w, Met1-Thr172. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 90316 | |
| Unconjugated | |
| BCL2L2 | |
| Apoptosis regulator Bcl-W; AW048834; BCL2 like 2; BCL2L2; BCL2-L-2; BCL2-like 2; BCL2-like 2 protein; Bcl-2-like protein 2; Bcl2-like protein 2; BCLW; BCL-W; BCL-WEL; BCL-WS; c98; Gtrgal2; Gtrosa41; Kiaa0271; PPP1R51; protein phosphatase 1, regulatory subunit 51 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 599 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction