missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCL-W/BCL2L2 Mouse anti-Human, Clone: CPTC-BCL2L2-2, Novus Biologicals™
Mouse Monoclonal Antibody
£196.00
Specifications
| Antigen | BCL-W/BCL2L2 |
|---|---|
| Clone | CPTC-BCL2L2-2 |
| Concentration | 0.2 mg/ml |
| Dilution | Western Blot 1-2 ug/ml, Protein Array |
| Applications | Western Blot, Peptide Array |
Description
BCL-W/BCL2L2 Monoclonal specifically detects BCL-W/BCL2L2 in Human samples. It is validated for Western Blot, Protein Array.Specifications
| BCL-W/BCL2L2 | |
| 0.2 mg/ml | |
| Western Blot, Peptide Array | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Biologically Active Proteins, Cancer | |
| 10 mM PBS with 0.05% BSA | |
| 599 | |
| IgG1 κ | |
| Protein A or G purified |
| CPTC-BCL2L2-2 | |
| Western Blot 1-2 ug/ml, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Apoptosis regulator Bcl-W, Bcl2-L-2, BCL2-like 2, BCL-W, BCLWbcl-2-like protein 2, KIAA0271BCL2-L-2 | |
| Recombinant human full-length BCL-W/BCL2L2 protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title