missing translation for 'onlineSavingsMsg'
Learn More

BCL-W/BCL2L2 Antibody (CPTC-BCL2L2-2), Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18323753 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18323753 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18323753 Supplier Novus Biologicals Supplier No. NBP308778JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

BCL-W/BCL2L2 Monoclonal antibody specifically detects BCL-W/BCL2L2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen BCL-W/BCL2L2
Applications Western Blot
Classification Monoclonal
Clone CPTC-BCL2L2-2
Conjugate Janelia Fluor 646
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias Apoptosis regulator Bcl-W, Bcl2-L-2, BCL2-like 2, BCL-W, BCLWbcl-2-like protein 2, KIAA0271BCL2-L-2
Host Species Mouse
Immunogen Recombinant human full-length BCL-W/BCL2L2 protein
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Biologically Active Proteins, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 599
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.