missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCL-W/BCL2L2 Antibody (003), Alexa Fluor™ 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89248AF405
This item is not returnable.
View return policy
Description
BCL-W/BCL2L2 Monoclonal antibody specifically detects BCL-W/BCL2L2 in Human samples. It is validated for ELISA
Specifications
| BCL-W/BCL2L2 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| Apoptosis regulator Bcl-W, Bcl2-L-2, BCL2-like 2, BCL-W, BCLWbcl-2-like protein 2, KIAA0271BCL2-L-2 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human BCL-W/BCL2L2 (Uniprot#: Q92843-1; Met1-Thr172). | |
| 0.1 mL | |
| Apoptosis, Biologically Active Proteins, Cancer | |
| 599 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 003 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction