missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bcl-rambo Polyclonal Antibody
Description
Bcl-rambo Polyclonal Antibody for Western Blot, IHC
Specifications
Specifications
| Antigen | Bcl-rambo |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | BCL2L13 |
| Gene Accession No. | P59017, Q9BXK5 |
| Gene Alias | BCL2 like 13; BCL2L13; Bcl2-L-13; BCL2-like 13 (apoptosis facilitator); bcl-2-like protein 13; Bcl-rambo; CD003; E430016C20Rik; MIL1; Mil-1; mitochondria located 1 homolog; protein Mil1 |
| Gene Symbols | BCL2L13 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?