missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bcl-G Recombinant Rabbit Monoclonal Antibody (K01_2L52)

Product Code. 30829357
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30829357 100 μL
30827345 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30829357 Supplier Invitrogen™ Supplier No. MA564006
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Bcl-G Recombinant Monoclonal Antibody for ICC/IF, Western Blot

The protein encoded by this gene belongs to the BCL2 protein family. BCL2 family members form hetero- or homodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. Overexpression of this gene has been shown to induce apoptosis in cells. Three alternatively spliced transcript variants encoding two distinct isoforms have been reported for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bcl-G
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone K01_2L52
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 150mM NaCl, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gene BCL2L14
Gene Accession No. Q6AYK4, Q9BZR8, Q9CPT0
Gene Alias 4930452K23Rik; 4933405K19Rik; 9030625M01Rik; AI429211; Apoptosis facilitator Bcl-2-like protein 14; apoptosis regulator BCL-G; BCL2 like 14; Bcl2l14; bcl2-L-14; BCL2-like 14 (apoptosis facilitator); BCLG; testicular tissue protein Li 26
Gene Symbols BCL2L14
Host Species Rabbit
Immunogen Recombinant protein of human Bcl G/BCL2L14.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 500348, 66813, 79370
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.