missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Bcl-G Recombinant Rabbit Monoclonal Antibody (24GB3005)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556994
This item is not returnable.
View return policy
Description
Bcl-G Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The protein encoded by this gene belongs to the BCL2 protein family. BCL2 family members form hetero- or homodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. Overexpression of this gene has been shown to induce apoptosis in cells. Three alternatively spliced transcript variants encoding two distinct isoforms have been reported for this gene.
Specifications
| Bcl-G | |
| Recombinant Monoclonal | |
| Unconjugated | |
| BCL2L14 | |
| 4930452K23Rik; 4933405K19Rik; 9030625M01Rik; AI429211; Apoptosis facilitator Bcl-2-like protein 14; apoptosis regulator BCL-G; BCL2 like 14; Bcl2l14; bcl2-L-14; BCL2-like 14 (apoptosis facilitator); BCLG; testicular tissue protein Li 26 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 500348, 66813, 79370 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB3005 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6AYK4, Q9BZR8, Q9CPT0 | |
| BCL2L14 | |
| A synthesized peptide derived from human BCLG (aa 1-250). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction