Learn More
Invitrogen™ Bcl-G Monoclonal Antibody (1D11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA522576
Description
Peptide Sequence: MCSTSGCDLE EIPLDDDDLN TIEFKILAYY TRHHVFKSTP ALFSPKLLRT RSLSQRGLGN CSANESWTEV SWPCRNSQSS EKAINLGKKK SSWKAFFGVV EKEDSQSTPA KVSAQGQRTL EYQDSHSQQW SRCLSNVEQC LEHEAVDPKV ISIANRVAEI VYSWPPPQAT QAGGFKSKEI FVTEGLSFQL QGHVPVASSS KKDEEEQILA KIVELLKYSG DQLERKLKKD KALMGHFQDG LSYSVFKTIT DQVLMGVDPR GESEVKAQGF KAALVIDVTA KLTAIDNHPM NRVLGFGTKY LKENFSPWIQ QHGGWEKILG ISHEEVD.
The protein encoded by this gene belongs to the BCL2 protein family. BCL2 family members form hetero- or homodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. Overexpression of this gene has been shown to induce apoptosis in cells. Three alternatively spliced transcript variants encoding two distinct isoforms have been reported for this gene.
Specifications
| Bcl-G | |
| Monoclonal | |
| 0.45 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q9BZR8 | |
| BCL2L14 | |
| BCL2L14 (AAH25778.1, 1 a.a. ~ 327 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Immunoprecipitation, Western Blot | |
| 1D11 | |
| Unconjugated | |
| BCL2L14 | |
| 4930452K23Rik; 4933405K19Rik; 9030625M01Rik; AI429211; Apoptosis facilitator Bcl-2-like protein 14; apoptosis regulator BCL-G; BCL2 like 14; Bcl2l14; bcl2-L-14; BCL2-like 14 (apoptosis facilitator); BCLG; testicular tissue protein Li 26 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 79370 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.