missing translation for 'onlineSavingsMsg'
Learn More

Bcl G Antibody (OTI6D1), DyLight 594, Novus Biologicals™

Product Code. 18410088 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18410088 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18410088 Supplier Novus Biologicals Supplier No. NBP272204DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Bcl G Monoclonal antibody specifically detects Bcl G in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bcl G
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI6D1
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias apoptosis facilitator Bcl-2-like protein 14, Apoptosis regulator Bcl-G, Bcl2-L-14, BCL2-like 14 (apoptosis facilitator), BCL-G, BCLGbcl2-L-14
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 116-216 of human Bcl G (NP_620048) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 79370
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.