missing translation for 'onlineSavingsMsg'
Learn More

Bcl G Antibody (1D11), Novus Biologicals™

Product Code. 18418258 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18418258 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18418258 Supplier Novus Biologicals Supplier No. H00079370M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Bcl G Monoclonal antibody specifically detects Bcl G in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation, Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bcl G
Applications Western Blot, ELISA, Immunoprecipitation, Sandwich ELISA
Classification Monoclonal
Clone 1D11
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunoprecipitation, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH25778
Gene Alias apoptosis facilitator Bcl-2-like protein 14, Apoptosis regulator Bcl-G, Bcl2-L-14, BCL2-like 14 (apoptosis facilitator), BCL-G, BCLGbcl2-L-14
Host Species Mouse
Immunogen BCL2L14 (AAH25778.1, 1 a.a. ∽ 327 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MCSTSGCDLEEIPLDDDDLNTIEFKILAYYTRHHVFKSTPALFSPKLLRTRSLSQRGLGNCSANESWTEVSWPCRNSQSSEKAINLGKKKSSWKAFFGVVEKEDSQSTPAKVSAQGQRTLEYQDSHSQQWSRCLSNVEQCLEHEAVDPKVISIANRVAEIVYSWPPPQATQAGGFKSKEIFVTEGLSFQLQGHVPVASSSKKDEEEQILAKIVELLKYSGDQLERKLKKDKALMGHFQDGLSYSVFKTITDQVLMGVDPRGESEVKAQGFKAALVIDVTAKLTAIDNHPMNRVLGFGTKYLKENFSPWIQQHGGWEKILGISHEEVD
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 79370
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.