missing translation for 'onlineSavingsMsg'
Learn More

Bcl-9 Antibody (2D4), Novus Biologicals™

Product Code. 18385328 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18385328 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18385328 Supplier Novus Biologicals Supplier No. H00000607M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Bcl-9 Monoclonal antibody specifically detects Bcl-9 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bcl-9
Applications Western Blot, ELISA, Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone 2D4
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_004317
Gene Alias B-cell CLL/lymphoma 9, B-cell CLL/lymphoma 9 protein, B-cell lymphoma 9 protein, bcl-9, LGS, MGC131591, Protein legless homolog
Host Species Mouse
Immunogen BCL9 (NP_004317, 1036 a.a. ~ 1135 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. DDDSPPARSPNLPSMNNMPGMGINTQNPRISGPNPVVPMPTLSPMGMTQPLSHSNQMPSPNAVGPNIPPHGVPMGPGLMSHNPIMGHGSQEPPMVPQGRM
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 607
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.