missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bcl-6 Mouse, Clone: BCL6/1527, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 604-MSM7-P0
This item is not returnable.
View return policy
Specifications
| Bcl-6 | |
| Nuclear | |
| BCL6/1527 | |
| Unconjugated | |
| NP_001124317 | |
| Mouse | |
| 95kDa | |
| Protein A/G Purified | |
| RUO | |
| 604 | |
| IgG1 κ |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| BCL6 | |
| Recombinant human bcl-6 protein fragment (aa256-389) | |
| Raji or Ramos cells. Tonsil or Hodgkin's lymphoma. | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction