missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bcl-6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-76541
This item is not returnable.
View return policy
Description
Bcl-6 Polyclonal specifically detects Bcl-6 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Bcl-6 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml | |
| B-cell CLL/lymphoma 6, B-cell lymphoma 6 protein transcript, BCL-5, BCL5B-cell lymphoma 5 protein, BCL-6, BCL6A, cys-his2 zinc finger transcription factor, LAZ3ZBTB27B-cell lymphoma 6 protein, lymphoma-associated zinc finger gene on chromosome 3, Protein LAZ-3, Zinc finger and BTB domain-containing protein 27, Zinc finger protein 51ZNF51, zinc finger transcription factor BCL6S | |
| Rabbit | |
| Affinity Purified | |
| Cancer, Cell Cycle and Replication, Tumor Suppressors | |
| 604.0 | |
| Human | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide | |
| BCL6 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: EHVVDTCRKFIKASEAEMVSAIKPPREEFLNSRMLMPQDIMAYRGREVVENNLPLRSAPGCESRAFAPSLYSGLSTPPASYSMYSHLPVSSLLFSDEEFRDVR | |
| 100 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction