Learn More
Bcl-6 Antibody (CL14611) [PerCP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26872PCP
Product Code. 30295264
  • £558.00 / 0.10mL
Description
Bcl-6 Monoclonal antibody specifically detects Bcl-6 in Human samples. It is validated for Immunohistochemistry,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Bcl-6
Monoclonal
PerCP
B-cell CLL/lymphoma 6, B-cell lymphoma 6 protein transcript, BCL-5, BCL5B-cell lymphoma 5 protein, BCL-6, BCL6A, cys-his2 zinc finger transcription factor, LAZ3ZBTB27B-cell lymphoma 6 protein, lymphoma-associated zinc finger gene on chromosome 3, Protein LAZ-3, Zinc finger and BTB domain-containing protein 27, Zinc finger protein 51ZNF51, zinc finger transcription factor BCL6S
This antibody was developed using a recombinant protein derived from P41182, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer, Cell Cycle and Replication, Tumor Suppressors
604
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Immunocytochemistry/Immunofluorescence
CL14611
PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified