missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bcl-6 Antibody (BCL6/1982), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-75749DL594
This item is not returnable.
View return policy
Description
Bcl-6 Monoclonal specifically detects Bcl-6 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Bcl-6 | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| BCL6 | |
| Recombinant human bcl-6 protein fragment (around aa256-389) (exact sequence is proprietary) (Uniprot: P41182) | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Tumor Suppressors | |
| Recognizes a protein of 95kDa, which is identified as Bcl-6. Antibody to bcl-6 is helpful in a number of diagnostic settings: (1) In the differential diagnosis of small B-cell lymphoma. Follicular lymphoma will show bcl-6 (and CD10) positivity whereas other small B-cell lymphomas are usually negative. (2) Bcl-6 is an important prognostic marker in diffuse large B-cell lymphomas (DLBCL), where CD10, bcl-6 and MUM1/IRF4 are used to identify germinal center and activated B-cell phenotypes. (3) Bcl-6 can be valuable in distinguishing classical Hodgkin lymphoma from nodular lymphocyte predominant Hodgkin lymphoma (NLPHL). The Reed-Sternberg cells of classical Hodgkin lymphoma are bcl-6 negative whereas the large ('LH') cells of NLPHL are bcl-6 positive. In contrast, anti-Bcl-6 rarely stains mantle-cell lymphoma and MALT lymphoma. | |
| Store at 4°C in the dark. |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| BCL6/1982 | |
| ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| B-cell CLL/lymphoma 6, B-cell lymphoma 6 protein transcript, BCL-5, BCL5B-cell lymphoma 5 protein, BCL-6, BCL6A, cys-his2 zinc finger transcription factor, LAZ3ZBTB27B-cell lymphoma 6 protein, lymphoma-associated zinc finger gene on chromosome 3, Protein LAZ-3, Zinc finger and BTB domain-containing protein 27, Zinc finger protein 51ZNF51, zinc finger transcription factor BCL6S | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction