missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Bcl-3 Monoclonal Antibody (Ham 150-3.5), Biotin

Armenian Hamster Monoclonal Antibody
£257.00 - £500.00
Specifications
| Antigen | Bcl-3 |
|---|---|
| Clone | Ham 150-3.5 |
| Concentration | 0.1 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | ELISA, Flow Cytometry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15323951
![]() |
Invitrogen™
MA517665 |
100 μg |
£257.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15333951
|
Invitrogen™
MA517666 |
300 μg |
£500.00
300µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Bcl-3 Monoclonal Antibody for Flow, ELISA
B cell Lymphoma 3 is a member of the IkB subfamily of inhibitors. This subfamily is part of the NF-kB transcription factor protein family and it is suggested that a balance in the concentration of various NF-kB family members regulates apoptosis and survival of activated T cells. Bcl-3 has been shown to have versatile functions such as cytoplasmic activation of p50 homodimers, translocation to the nucleus, and modulation of the transcriptional machinery in the nucleus. Bcl-3 activity often relies on several nuclear interacting proteins such as Tip60, Jab1, Bard1, and Pirin.Specifications
| Bcl-3 | |
| 0.1 mg/mL | |
| ELISA, Flow Cytometry | |
| Biotin | |
| Armenian Hamster | |
| RUO | |
| PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
| Q9Z2F6 | |
| 12051 | |
| recombinant Bcl-3-6-histidine tag. | |
| Antibody |
| Ham 150-3.5 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Monoclonal | |
| Liquid | |
| IgG | |
| Mouse | |
| Bcl3 | |
| AI528691; B cell leukemia/lymphoma 3; B-cell CLL/lymphoma 3; B-cell leukemia/lymphoma 3; B-cell lymphoma 3 protein; B-cell lymphoma 3 protein homolog; B-cell lymphoma 3-encoded protein; BCL3; BCL-3; Bcl-3 protein; BCL4; chronic lymphatic leukemia protein; D19S37; Proto-oncogene BCL3 | |
| Bcl3 | |
| Primary | |
| Protein G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title