missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bcl-2 related protein A1 Antibody (OTI3D10), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70246C
This item is not returnable.
View return policy
Description
Bcl-2 related protein A1 Monoclonal specifically detects Bcl-2 related protein A1 in Human samples. It is validated for Western Blot.
Specifications
| Bcl-2 related protein A1 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| BCL2A1 | |
| Full length recombinant protein of human Bcl-2 related protein A1 (NP_004040) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot | |
| OTI3D10 | |
| Western Blot | |
| ACC-1, ACC-2, Bcl2-L-5, BCL2L5HBPA1, Bcl-2-like protein 5, BCL2-related protein A1, BFL1bcl-2-related protein A1, GRSbcl2-L-5, hematopoietic BCL2-related protein A1, Hemopoietic-specific early response protein, Protein BFL-1, Protein GRS | |
| Mouse | |
| Affinity Purified | |
| Apoptosis, Cancer | |
| 597.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction