missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bcl-2 related protein A1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP1-76715-0.025mg
This item is not returnable.
View return policy
Description
Bcl-2 related protein A1 Polyclonal specifically detects Bcl-2 related protein A1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Bcl-2 related protein A1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| ACC-1, ACC-2, Bcl2-L-5, BCL2L5HBPA1, Bcl-2-like protein 5, BCL2-related protein A1, BFL1bcl-2-related protein A1, GRSbcl2-L-5, hematopoietic BCL2-related protein A1, Hemopoietic-specific early response protein, Protein BFL-1, Protein GRS | |
| Rabbit | |
| 20 kDa | |
| 0.025 mg | |
| Apoptosis, Cancer | |
| 597 | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 10 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 10 μg/mL | |
| NP_004040 | |
| BCL2A1 | |
| Antibody was raised against a 14 amino acid synthetic peptide from near the carboxy terminus of human Bfl-1. The immunogen is located within the last 50 amino acids of Bfl-1. Amino Acid Squence: ENGFVKKFEPKSG | |
| Affinity Purified | |
| RUO | |
| Primary | |
| At least two isoforms of Bfl-1 are known to exist; this antibody will only detect the larger isoform. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction