Learn More
Bcl-2 Mouse anti-Human, DyLight 755, Clone: 100/D5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-33315IR
 View more versions of this product
Product Code. 18618699
  • £462.00 / 0.10mL
Description
Bcl-2 Monoclonal specifically detects Bcl-2 in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Bcl-2
Monoclonal
DyLight 755
50 mM Sodium Borate
Mouse
Protein G purified
RUO
Primary
Human, Mouse (Negative), Rat (Negative)
Purified
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
100/D5
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1-5μg/mL for coating, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
apoptosis regulator Bcl-2, B-cell CLL/lymphoma 2, Bcl-2
The synthetic peptide corresponding to amino acids range from 41-64 (GAAPAPGIFSSQPG-Cys), of human Bcl-2 was used as immunogen to generate the antibody; GenBank no. NP_000648.2 (Pezzella et al, 1990).
0.1 mL
Apoptosis, Autophagy, Cancer, Cellular Markers, Phospho Specific, Tumor Suppressors
596
Store at 4°C in the dark.
IgG1 κ