Learn More
Invitrogen™ Bcl-2 Monoclonal Antibody (Bcl-2/100), eBioscience™

Mouse Monoclonal Antibody
£274.00 - £1376.00
Specifications
| Antigen | Bcl-2 |
|---|---|
| Clone | Bcl-2/100 |
| Concentration | 0.5 mg/mL |
| Content And Storage | 4°C |
| Applications | Immunohistochemistry, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17120542
|
Invitrogen™
14-1028-37 |
2 mg |
£1376.00
2mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15287137
![]() |
Invitrogen™
14-1028-82 |
100 μg |
£274.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Description: This bcl-2/100 monoclonal antibody reacts to human Bcl-2 (B-cell Lymphoma 2), the founding member of a family of pro-apoptotic proteins which includes Bcl-XL, Bcl-w, and Mcl-1. Bcl-2 plays a role in protecting against apoptosis-inducing insults such as dexamethasone and staurosporine, as well as irradiation and cytokine withdrawal. The mechanism is thought to include regulation of mitochondrial membrane permeability, resulting in sequestration of caspase proforms, or by preventing the release of apoptosis-mediating mitochondrial factors, such as cytochrome c and AIF (apoptosis-inducing factor), into the cytoplasm. Bcl-2 is localized to the cytoplasmic side of the mitochondrial membrane, the nuclear envelope, and the endoplasmic reticulum. Interestingly Bcl-2 does not play a role in preventing cell death of autoreactive T cells in the thymus or Fas-mediated apoptosis. Two isoforms, termed alpha and beta of Bcl-2 have been identified and are a result of alternative splicing at the C-terminus. This antibody recognizes both forms. Applications Reported: This Bcl-2/100 antibody has been reported for use in immunoblotting (WB). Applications Tested: This Bcl-2/100 antibody has been tested by immunoblot analysis of the Jurkat cell line. This can be used at 1-5 μg/mL for detection of reduced and denatured protein. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
BCL-2 is a key regulator of apoptosis that functions to either inhibit or promote cell death. The BCL-2 family members are also characterized by dimerizing to further modulate apoptosis. Bag1, for example, has been found to form a heterodimer with BCL-2 resulting in the enhancement of the anti-apoptotic effect of BCL-2. Bax and Bak have been shown to play a critical role in cytochrome c release from mitochondria and thus initiate apoptosis. Bax exerts a pro-apoptotic rather than an anti-apoptotic effect on cells. Constitutive expression of BCL2, such as in the case of translocation of BCL2 to Ig heavy chain locus, is thought to be the cause of follicular lymphoma. In most follicular lymphomas, neoplastic germinal centers express high levels of BCL-2 alpha protein, whereas the normal or hyperplastic germinal centers are negative. Two transcript variants of BCL-2, produced by alternate splicing, differ in their C-terminal ends. The overexpression of BCL-2 has been linked to human cancers such as B-cell lymphoma and prostate cancer.Specifications
| Bcl-2 | |
| 0.5 mg/mL | |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| P10415 | |
| 596 | |
| Primary | |
| Affinity chromatography |
| Bcl-2/100 | |
| 4°C | |
| Monoclonal | |
| Liquid | |
| IgG1 κ | |
| Human | |
| Bcl2 | |
| apoptosis regulator Bcl-2; AW986256; B cell leukemia/lymphoma 2; B cell lymphoma 2 associated oncogene; B-cell CLL/lymphoma 2; B-cell leukemia/lymphoma 2; Bcl 2; BCL2; bcl-2; BCL2 apoptosis regulator; Bcl-2 protein; BCL2, apoptosis regulator; Bcl2-like protein; C430015F12Rik; D630044D05Rik; D830018M01Rik; OTTHUMP00000163680; PPP1R50; protein phosphatase 1, regulatory subunit 50 | |
| Bcl2 | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.