missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bcl-2 Antibody (BCL2/2210R), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-75761-20ug
This item is not returnable.
View return policy
Description
Bcl-2 Monoclonal antibody specifically detects Bcl-2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array
Specifications
| Bcl-2 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1-2 μg/mL, Immunohistochemistry, Immunohistochemistry-Paraffin 1-2 μg/mL, Protein Array | |
| P10415, P10415, P10415, P10415 | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array | |
| BCL2/2210R | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| apoptosis regulator Bcl-2, B-cell CLL/lymphoma 2, Bcl-2 | |
| Recombinant full-length human bcl-2 protein (Uniprot: P10415) | |
| 20 μg | |
| Apoptosis, Autophagy, Biologically Active Proteins, Cancer, Cellular Markers, Mitochondrial Mediated Pathway, Oncogenes, Tumor Suppressors | |
| 596 | |
| Store at 4°C. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction