missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Bcl-2 Monoclonal antibody specifically detects Bcl-2 in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
Specifications
Specifications
| Antigen | Bcl-2 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), CyTOF |
| Classification | Monoclonal |
| Clone | 100/D5+124 |
| Conjugate | PerCP |
| Formulation | PBS |
| Gene Alias | apoptosis regulator Bcl-2, B-cell CLL/lymphoma 2, Bcl-2 |
| Host Species | Mouse |
| Immunogen | A synthetic peptide, aa41-54 (GAAPAPGIFSSQPG-Cys) of human Bcl-2 (100/D5 & 124) (Uniprot: P10415) |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?