missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BCL-10 Recombinant Rabbit Monoclonal Antibody (23GB1210), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553201
This item is not returnable.
View return policy
Description
BCL-10 is an N-terminal CARD (Caspase Recruitment Domain) containing protein that is involved in the adaptive immune response. It promotes apoptosis, pro-caspase-9 maturation, and activation of NF-kappa-B via NIK and IKK. BCL-10 is also an adapter protein between upstream TNFR1-TRADD-RIP complex and the downstream NIK-IKK-IKAP complex. It is a substrate for MALT1. Mutations in the gene can lead to Immmunodeficiency 37 or Lymphoma, mucosa-associated lymphoid type.
Specifications
| BCL-10 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| BCL10 | |
| AI132454; B cell CLL/lymphoma 10; B cell leukemia/lymphoma 10; B-cell CLL/lymphoma 10; B-cell leukemia/lymphoma 10; B-cell lymphoma/leukemia 10; BCL10; Bcl-10; BCL10 immune signaling adaptor; C81403; CARD containing molecule enhancing NF-kB; CARD-containing apoptotic signaling protein; CARD-containing molecule enhancing NF-kappa-B; CARD-containing proapoptotic protein; CARD-like apoptotic protein; CARMEN; caspase-recruiting domain-containing protein; cCARMEN; cE10; c-E10; CED-3/ICH-1 prodomain homologous E10-like regulator; cellular homolog of vCARMEN; Cellular-E10; CIPER; Clap; hCLAP; IMD37; Mamm; mammalian CARD-containing adapter molecule E10; mCIPER; mCLAP; mE10; OTTHUMP00000011647; OTTHUMP00000036080; RCD; RP11-234D19.2; R-RCD1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 8915 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB1210 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O95999 | |
| BCL10 | |
| A synthesized peptide derived from human Bcl10 (1-233AA). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction