missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bcl-10 Polyclonal Antibody

Bcl-10 Polyclonal Antibody
Brand: Invitrogen PA524191
This item is not returnable.
View return policy
Description
Bcl-10 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Specifications
| Bcl-10 | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| O95999, Q9Z0H7 | |
| BCL10 | |
| KLH conjugated synthetic peptide between 24-51 amino acids from the N-terminal region of human BCL10. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| BCL10 | |
| AI132454; B cell CLL/lymphoma 10; B cell leukemia/lymphoma 10; B-cell CLL/lymphoma 10; B-cell leukemia/lymphoma 10; B-cell lymphoma/leukemia 10; BCL10; Bcl-10; BCL10 immune signaling adaptor; C81403; CARD containing molecule enhancing NF-kB; CARD-containing apoptotic signaling protein; CARD-containing molecule enhancing NF-kappa-B; CARD-containing proapoptotic protein; CARD-like apoptotic protein; CARMEN; caspase-recruiting domain-containing protein; cCARMEN; cE10; c-E10; CED-3/ICH-1 prodomain homologous E10-like regulator; cellular homolog of vCARMEN; Cellular-E10; CIPER; Clap; hCLAP; IMD37; Mamm; mammalian CARD-containing adapter molecule E10; mCIPER; mCLAP; mE10; OTTHUMP00000011647; OTTHUMP00000036080; RCD; RP11-234D19.2; R-RCD1 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 12042, 8915 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction